(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Ser-OMe·HCl, H-D-Ser-OMe.HCl, D-Serine methyl ester hydrochloride

A128866-100G A128866-10G A128866-1G A128866-500G A128866-25G
76822-864EA 41 USD
76822-864 76822-866 76822-868 76822-870 77313-546
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉O₃N·HCl
MW: 155.6 g/mol
MDL Number: MFCD00066121
CAS Number: 5874-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR