5-Aminoisophthalic acid 98%

Supplier: Ambeed
Warning

A128851-1000G A128851-100G A128851-10G A128851-25G A128851-500G
76822-856EA 73 USD
76822-856 76822-858 76822-860 76822-862 77611-136
5-Aminoisophthalic acid 98%
5-Aminoisophthalic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00002522
CAS Number: 99-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR