2-Amino-9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one 97%

Supplier: Ambeed
Warning

A128554-100MG A128554-1G A128554-250MG A128554-5G
76822-848EA 41.38 USD
76822-848 76822-850 76822-852 76822-854
2-Amino-9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one 97%
2-Amino-9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₉N₅O₆
MW: 341.32 g/mol
MDL Number: MFCD02682962
CAS Number: 473278-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR