Methyl 5-bromo-2-cyanobenzoate 95%

Supplier: Ambeed
Warning

A128523-100MG A128523-1G A128523-250MG A128523-5G
76822-840EA 28.65 USD
76822-840 76822-842 76822-844 76822-846
Methyl 5-bromo-2-cyanobenzoate 95%
Methyl 5-bromo-2-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.05 g/mol
MDL Number: MFCD15523586
CAS Number: 714237-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR