2-Dimethylphosphorylaniline 98%

Supplier: Ambeed
Warning

Synonyms: (2-Aminophenyl)dimethylphosphine oxide

A127667-100MG A127667-10G A127667-1G A127667-250MG A127667-5G A127667-25G
76822-800EA 6.37 USD
76822-800 76822-802 76822-804 76822-806 76822-808 77683-131
2-Dimethylphosphorylaniline 98%
2-Dimethylphosphorylaniline

Precaución: For research use only.

Formula: C₈H₁₂NOP
MW: 169.161 g/mol
MDL Number: MFCD23135529
CAS Number: 1197953-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR