2-Methylbenzo[d]thiazole-5-carbonitrile 97%

Supplier: Ambeed
Warning

A127333-100MG A127333-1G A127333-250MG
76822-772EA 83.82 USD
76822-772 76822-774 76822-776
2-Methylbenzo[d]thiazole-5-carbonitrile 97%
2-Methylbenzo[d]thiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂S
MW: 174.22 g/mol
MDL Number: MFCD18415973
CAS Number: 90418-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR