(4-Amino-6-chloropyridin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A127319-100MG A127319-1G A127319-250MG
76822-766EA 44.56 USD
76822-766 76822-768 76822-770
(4-Amino-6-chloropyridin-3-yl)methanol 95%
(4-Amino-6-chloropyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.59 g/mol
MDL Number: MFCD18914390
CAS Number: 846036-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR