3'-Bromo-2'-methoxyacetophenone 95%

Supplier: Ambeed
Warning

A127027-100MG A127027-1G A127027-250MG
76822-760EA 10 USD
76822-760 76822-762 76822-764
3'-Bromo-2'-methoxyacetophenone 95%
3'-Bromo-2'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD22573042
CAS Number: 267651-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR