4-Chloro-1H-indol-7-amine 97%

Supplier: Ambeed
Warning

A126852-1G A126852-250MG A126852-100MG
76822-752EA 247 USD
76822-752 76822-754 77885-126
4-Chloro-1H-indol-7-amine 97%
4-Chloro-1H-indol-7-amine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MDL Number: MFCD11007923
CAS Number: 292636-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR