Ethyl 4-methyl-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A126742-100MG A126742-1G A126742-250MG
76822-742EA 22.28 USD
76822-742 76822-744 76822-746
Ethyl 4-methyl-1H-indole-2-carboxylate 97%
Ethyl 4-methyl-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
MDL Number: MFCD00981705
CAS Number: 16732-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR