(S)-5-Phenyl-1H-pyrrol-3-yl 2-(4-methylphenylsulfonamido)propanoate 95%

Supplier: Ambeed
Warning

A126641-100MG A126641-1G A126641-250MG
76822-736EA 31.83 USD
76822-736 76822-738 76822-740
(S)-5-Phenyl-1H-pyrrol-3-yl 2-(4-methylphenylsulfonamido)propanoate 95%
(S)-5-Phenyl-1H-pyrrol-3-yl 2-(4-methylphenylsulfonamido)propanoate

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₄S
MW: 384.45 g/mol
MDL Number: MFCD08704359
CAS Number: 99740-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR