N-Me-Ser-OH.HCl 95%

Supplier: Ambeed
Warning

A126639-1G A126639-250MG A126639-5G
76822-730EA 117 USD
76822-730 76822-732 76822-734
N-Me-Ser-OH.HCl 95%
N-Me-Ser-OH.HCl

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO₃
MW: 155.58 g/mol
MDL Number: MFCD06795803
CAS Number: 141193-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR