2-(Chloromethyl)-3-fluoropyridine hydrochloride 95%

Supplier: Ambeed
Danger

A126478-100MG A126478-250MG
76822-720EA 27.59 USD
76822-720 77683-124
2-(Chloromethyl)-3-fluoropyridine hydrochloride 95%
2-(Chloromethyl)-3-fluoropyridine hydrochloride

Precaución: For research use only.

Formula: C₆H₆Cl₂FN
MW: 182.02 g/mol
MDL Number: MFCD19442768
CAS Number: 149463-07-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR