2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A125628-10G A125628-25G A125628-5G A125628-1G
76822-698EA 169 USD
76822-698 76822-700 76822-702 77571-530
2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline hydrochloride 95%
2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClN
MDL Number: MFCD00193051
CAS Number: 4071-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR