(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Lys(Mmt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-Methoxyphenyl)diphenylmethyl)amino)hexanoic acid, Fmoc-Lys(4-methoxytrityl)-OH

A125481-1G A125481-250MG A125481-25G A125481-5G A125481-10G A125481-100G
76822-670EA 39.26 USD
76822-670 76822-672 76822-674 76822-676 77566-414 77683-118
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid

Precaución: For research use only.

Formula: C₄₁H₄₀N₂O₅
MW: 640.78 g/mol
MDL Number: MFCD00270542
CAS Number: 159857-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR