3-(2-Cyanopropan-2-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A125231-250MG A125231-5G A125231-1G
76822-660EA 16.98 USD
76822-660 76822-662 77592-430
3-(2-Cyanopropan-2-yl)benzoic acid 98%
3-(2-Cyanopropan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
MDL Number: MFCD19694856
CAS Number: 872091-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR