(R)-(3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl methanesulfonate 97%

Supplier: Ambeed
Warning

A124997-100G A124997-10G A124997-1G A124997-25G A124997-5G
76822-642EA 230.22 USD
76822-642 76822-644 76822-646 76822-648 76822-650
(R)-(3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl methanesulfonate 97%
(R)-(3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl methanesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₁₉FN₂O₆S
MW: 374.39 g/mol
MDL Number: MFCD18070719
CAS Number: 174649-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR