1-Chloro-6-fluoroisoquinoline 97%

Supplier: Ambeed
Warning

A124965-100MG A124965-1G A124965-250MG
76822-636EA 30.77 USD
76822-636 76822-638 76822-640
1-Chloro-6-fluoroisoquinoline 97%
1-Chloro-6-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClFN
MW: 181.594 g/mol
MDL Number: MFCD09959717
CAS Number: 214045-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR