N,N-Dimethylglycine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethylglycine HCl

A124936-100G A124936-25G A124936-500G A124936-5G A124936-1000G
76822-628EA 16.98 USD
76822-628 76822-630 76822-632 76822-634 77683-111
N,N-Dimethylglycine hydrochloride 98%
N,N-Dimethylglycine hydrochloride

Precaución: For research use only.

Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD00012610
CAS Number: 2491-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR