1-Propylpyrazole-4-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1-Propyl-1H-pyrazol-4-yl)boronic acid

A124755-1G A124755-250MG A124755-5G
76822-624EA 46 USD
76822-624 76822-626 77885-112
1-Propylpyrazole-4-boronic acid 97%
1-Propylpyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₆H₁₁BN₂O₂
MDL Number: MFCD11504862
CAS Number: 847818-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR