Methyl cinnoline-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A124558-100MG
A124558-1G
A124558-250MG
76822-612EA
149
USD
76822-612
76822-614
76822-616
Methyl cinnoline-6-carboxylate 95%
Methyl cinnoline-6-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₂ |
MDL Number:
MFCD20528017 CAS Number: 318276-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...