5-Chloro-4-iodo-2-nitroaniline 95%

Supplier: Ambeed
Warning

A124187-10G A124187-1G
76822-610EA 202.64 USD
76822-610 77683-108
5-Chloro-4-iodo-2-nitroaniline 95%
5-Chloro-4-iodo-2-nitroaniline

Precaución: For research use only.

Formula: C₆H₄ClIN₂O₂
MW: 298.47 g/mol
MDL Number: MFCD14706094
CAS Number: 335349-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR