Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A124133-100MG A124133-1G A124133-250MG A124133-5G
76822-604EA 44 USD
76822-604 76822-606 76822-608 77592-418
Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride 97%
Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₈ClNO₂
MDL Number: MFCD09839322
CAS Number: 135908-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR