Methyl 4-(4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridin-2-yl)benzoate 95%

Supplier: Ambeed
Warning

A123832-100MG A123832-250MG
76822-586EA 161.26 USD
76822-586 76822-588
Methyl 4-(4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridin-2-yl)benzoate 95%
Methyl 4-(4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridin-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₂₁H₁₆N₄O₂
MW: 356.38 g/mol
MDL Number: MFCD23135526
CAS Number: 886444-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR