2-Chloro-6-(trifluoromethyl)nicotinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-trifluoromethyl-pyridine-3-carbaldehyde

A123791-1G A123791-250MG A123791-5G
76822-570EA 120.95 USD
76822-570 76822-572 76822-574
2-Chloro-6-(trifluoromethyl)nicotinaldehyde 97%
2-Chloro-6-(trifluoromethyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO
MW: 209.553 g/mol
MDL Number: MFCD09907845
CAS Number: 944900-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR