Ethyl 4-oxopyrrolidine-3-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A123732-10G
A123732-1G
A123732-250MG
A123732-5G
76822-562EA
803.11
USD
76822-562
76822-564
76822-566
76822-568
Ethyl 4-oxopyrrolidine-3-carboxylate hydrochloride 95%
Ethyl 4-oxopyrrolidine-3-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₂ClNO₃ MW: 193.63 g/mol |
MDL Number:
MFCD17012780 CAS Number: 916814-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...