1,3,5-Trichloro-2-fluorobenzene 97%

Supplier: Ambeed
Warning

A123698-1G A123698-250MG A123698-5G
76822-558EA 24 USD
76822-558 76822-560 77885-106
1,3,5-Trichloro-2-fluorobenzene 97%
1,3,5-Trichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₃F
MDL Number: MFCD00060654
CAS Number: 36556-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR