(S)-Ethyl 3-(4-aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 95%

Supplier: Ambeed
Warning

A123218-100MG A123218-1G A123218-250MG
76822-532EA 20.16 USD
76822-532 76822-534 76822-536
(S)-Ethyl 3-(4-aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 95%
(S)-Ethyl 3-(4-aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₄
MW: 308.37 g/mol
MDL Number: MFCD18207036
CAS Number: 67630-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR