Tri-tert-Butyl 1,4,7,10-tetraazacyclododecane-1,4,7-tricarboxylate 96%

Supplier: Ambeed
Warning

A122617-250MG A122617-1G A122617-5G
76822-520EA 104 USD
76822-520 77592-388 77611-050
Tri-tert-Butyl 1,4,7,10-tetraazacyclododecane-1,4,7-tricarboxylate 96%
Tri-tert-Butyl 1,4,7,10-tetraazacyclododecane-1,4,7-tricarboxylate

Precaución: for Research Use Only

Formula: C₂₃H₄₄N₄O₆
MDL Number: MFCD09953471
CAS Number: 175854-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR