7-Bromo-3-chloroquinoline 98%

Supplier: Ambeed
Warning

A121672-100MG A121672-1G
76822-472EA 101 USD
76822-472 76822-474
7-Bromo-3-chloroquinoline 98%
7-Bromo-3-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD18253967
CAS Number: 1246549-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR