4-(5-Bromo-2-chlorobenzyl)phenol 97%

Supplier: Ambeed
Warning

A121591-1G A121591-250MG A121591-25G A121591-5G A121591-10G
76822-450EA 8.49 USD
76822-450 76822-452 76822-454 76822-456 77683-092
4-(5-Bromo-2-chlorobenzyl)phenol 97%
4-(5-Bromo-2-chlorobenzyl)phenol

Precaución: For research use only.

Formula: C₁₃H₁₀BrClO
MW: 297.58 g/mol
MDL Number: MFCD19441145
CAS Number: 864070-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR