Ethyl 3-amino-6-bromobenzofuran-2-carboxylate 95%

Supplier: Ambeed
Warning

A121512-100MG A121512-1G A121512-250MG
76822-438EA 29.71 USD
76822-438 76822-440 76822-442
Ethyl 3-amino-6-bromobenzofuran-2-carboxylate 95%
Ethyl 3-amino-6-bromobenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₃
MW: 284.106 g/mol
MDL Number: MFCD21609331
CAS Number: 887250-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR