3’-Bromo-2’,4’-difluoroacetophenone 98%

Supplier: Ambeed
Warning

A121272-250MG A121272-5G A121272-1G A121272-25G
76822-430EA 5 USD
76822-430 76822-432 77885-100 77885-101
3’-Bromo-2’,4’-difluoroacetophenone 98%
3’-Bromo-2’,4’-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MDL Number: MFCD13689251
CAS Number: 1210824-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR