1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Fluorophenyl)cyclobutanecarboxylic acid

A120872-1G A120872-250MG
76822-414EA 118.83 USD
76822-414 76822-416
1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid 98%
1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₂
MW: 194.202 g/mol
MDL Number: MFCD08444832
CAS Number: 179411-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR