tert-Butyl (3-mercaptopyridin-4-yl)carbamate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A120826-100MG A120826-250MG A120826-1G
76822-410EA 108.22 USD
76822-410 76822-412 77683-084
tert-Butyl (3-mercaptopyridin-4-yl)carbamate 95%, Ambeed.Inc
tert-Butyl (3-mercaptopyridin-4-yl)carbamate

Precaución: For research use only.

Formula: C₁₀H₁₄N₂O₂S
MW: 226.29 g/mol
MDL Number: MFCD21648454
CAS Number: 365996-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR