9-Bromo-[1,2,4]triazolo[1,5-c]quinazolin-5(6H)-one 95%

Supplier: Ambeed
Warning

A120522-100MG A120522-1G A120522-250MG
76822-380EA 66 USD
76822-380 76822-382 76822-384
9-Bromo-[1,2,4]triazolo[1,5-c]quinazolin-5(6H)-one 95%
9-Bromo-[1,2,4]triazolo[1,5-c]quinazolin-5(6H)-one

Precaución: for Research Use Only

Formula: C₉H₅BrN₄O
MDL Number: MFCD14584721
CAS Number: 882517-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR