4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A120456-100MG A120456-250MG
76822-376EA 39 USD
76822-376 76822-378
4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride 95%
4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂·ClH
MW: 205.682 g/mol
MDL Number: MFCD23701423
CAS Number: 854214-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR