(3-Bromo-2,5-dimethylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2,5-dimethylphenylboronic acid

A120328-100MG A120328-250MG
76822-362EA 10.61 USD
76822-362 76822-364
(3-Bromo-2,5-dimethylphenyl)boronic acid 97%
(3-Bromo-2,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₂
MW: 228.88 g/mol
MDL Number: MFCD12026703
CAS Number: 1259318-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR