4-(Difluoromethoxy)phenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Difluoromethoxy)phenyl)acetic acid

A119896-1G A119896-250MG A119896-100MG
76822-346EA 44 USD
76822-346 76822-348 77885-088
4-(Difluoromethoxy)phenylacetic acid 97%
4-(Difluoromethoxy)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.1600 g/mol
MDL Number: MFCD00236239
CAS Number: 243659-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR