Ethyl 5-chloro-4-methylthiophene-2-carboxylate 95+%

Supplier: Ambeed
Warning

A119789-1G A119789-250MG
76822-342EA 299.18 USD
76822-342 76822-344
Ethyl 5-chloro-4-methylthiophene-2-carboxylate 95+%
Ethyl 5-chloro-4-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClO₂S
MW: 204.67 g/mol
MDL Number: MFCD19053224
CAS Number: 74598-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR