(S)-4-(4-Hydroxybenzyl)oxazolidine-2,5-dione 97%

Supplier: Ambeed
Warning

A119438-1G A119438-5G
76822-326EA 20.16 USD
76822-326 76822-328
(S)-4-(4-Hydroxybenzyl)oxazolidine-2,5-dione 97%
(S)-4-(4-Hydroxybenzyl)oxazolidine-2,5-dione

Precaución: for Research Use Only

Formula: C₁₀H₉NO₄
MW: 207.18 g/mol
MDL Number: MFCD11046653
CAS Number: 3415-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR