(S)-tert-Butyl 2-(bromomethyl)morpholine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A119260-100MG
A119260-250MG
A119260-5G
A119260-1G
76822-318EA
17
USD
76822-318
76822-320
77571-144
77885-085
(S)-tert-Butyl 2-(bromomethyl)morpholine-4-carboxylate 97%
(S)-tert-Butyl 2-(bromomethyl)morpholine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈BrNO₃ |
MDL Number:
MFCD20489339 CAS Number: 919286-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...