1-(2-Bromo-ethyl)-piperidine hydrobromide 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromoethyl)piperidine hydrobromide

A119093-100MG A119093-250MG A119093-5G
76822-312EA 38.2 USD
76822-312 76822-314 76822-316
1-(2-Bromo-ethyl)-piperidine hydrobromide 97%
1-(2-Bromo-ethyl)-piperidine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₁₄BrN·BrH
MW: 273.009 g/mol
MDL Number: MFCD00463352
CAS Number: 89796-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR