(1S,2S)-2-(Benzylamino)cyclohexanol 97%

Supplier: Ambeed
Warning

A118820-10G A118820-5G
76822-290EA 50.93 USD
76822-290 76822-292
(1S,2S)-2-(Benzylamino)cyclohexanol 97%
(1S,2S)-2-(Benzylamino)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO
MW: 205.29 g/mol
MDL Number: MFCD10566419
CAS Number: 322407-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR