3-(Methoxymethyl)-morpholine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Methoxymethyl)morpholine hydrochloride

A118495-100MG A118495-1G
76822-282EA 66.84 USD
76822-282 76822-284
3-(Methoxymethyl)-morpholine hydrochloride 95%
3-(Methoxymethyl)-morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
MDL Number: MFCD18425620
CAS Number: 955400-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR