Ethyl 3-amino-5-chloropicolinate 95%

Supplier: Ambeed
Warning

A118489-100MG A118489-250MG A118489-1G
76822-278EA 42.44 USD
76822-278 76822-280 77683-067
Ethyl 3-amino-5-chloropicolinate 95%
Ethyl 3-amino-5-chloropicolinate

Precaución: For research use only.

Formula: C₈H₉ClN₂O₂
MW: 200.62 g/mol
MDL Number: MFCD10566235
CAS Number: 27330-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR