2-(Benzyloxy)-5-bromoanisole 97%

Supplier: Ambeed
Warning

A118207-10G A118207-1G A118207-250MG A118207-25G A118207-5G
76822-254EA 159 USD
76822-254 76822-256 76822-258 76822-260 76822-262
2-(Benzyloxy)-5-bromoanisole 97%
2-(Benzyloxy)-5-bromoanisole

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MDL Number: MFCD00092521
CAS Number: 63057-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR