(5-Chloro-2-propoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A117641-1G A117641-250MG
76822-212EA 16.98 USD
76822-212 76822-214
(5-Chloro-2-propoxyphenyl)boronic acid 95%
(5-Chloro-2-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
MW: 214.45 g/mol
MDL Number: MFCD06411348
CAS Number: 849062-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR