2-Methylimidazo[1,2-b]pyridazin-6-amine 97%
Supplier: Ambeed
|
Warning
|
A117445-10G
A117445-1G
A117445-250MG
A117445-25G
A117445-5G
76822-190EA
741
USD
76822-190
76822-192
76822-194
76822-196
76822-198
2-Methylimidazo[1,2-b]pyridazin-6-amine 97%
2-Methylimidazo[1,2-b]pyridazin-6-amine
Precaución: for Research Use Only
Formula:
C₇H₈N₄ |
MDL Number:
MFCD19211982 CAS Number: 154704-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...