Sodium-N-methyltaurinate 60% in water

Supplier: Ambeed
Warning

A117322-100G A117322-25G A117322-500G
76822-182EA 114.58 USD
76822-182 76822-184 76822-186 76822-188
Sodium-N-methyltaurinate 60% in water
Sodium-N-methyltaurinate

Precaución: for Research Use Only

Formula: C₃H₈NNaO₃S
MDL Number: MFCD00066598
CAS Number: 4316-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR